accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOOHNH2HCl

CAS RN 16012-55-8

IUPAC Name: 2-amino-3-(methylamino)propanoic acid hydrochloride
Synonyms: 3-(methylamino)-(DL)-alanine hydrochloride
Molecular Weight (g/mol): 154.59
Molecular Formula: C4H11ClN2O2
InChi Key: VDXYGASOGLSIDM-UHFFFAOYNA-N
SMILES: Cl.CNCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results