accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16015-71-7

IUPAC Name: 1-(3-methoxyphenyl)piperazine
Synonyms: 1-(3-methoxyphenyl)piperazine
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: PZIBVWUXWNYTNL-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)N1CCNCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)