accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160278-20-6

IUPAC Name: [4-(thiophen-3-yl)phenyl]methanol
Synonyms: [4-(thiophen-3-yl)phenyl]methanol
Molecular Weight (g/mol): 190.26
Molecular Formula: C11H10OS
InChi Key: KEHJEXSFOWCTHM-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results