accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrCH3

CAS RN 160313-69-9

IUPAC Name: ethyl 4-bromo-3-methylbenzoate
Synonyms: ethyl 4-bromo-3-methylbenzoate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: GIGDAWLJINYIFV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C)=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results