accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16034-40-5

IUPAC Name: 4-methoxy-N,N-diphenylbenzamide
Synonyms: 4-methoxy-N,N-diphenylbenzamide
Molecular Weight (g/mol): 303.36
Molecular Formula: C20H17NO2
InChi Key: GVALSXYCLDABKT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results