CAS RN 160388-54-5
CAS RN 160388-54-5
IUPAC Name:
4-[(1H-1,2,4-triazol-1-yl)methyl]benzoic acid
Synonyms:
4-(1,2,4-triazol-1-ylmethyl)benzoic acid
Molecular Weight (g/mol):
203.2
Molecular Formula:
C10H9N3O2
InChi Key:
DJXUEFGFCMIONN-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(CN2C=NC=N2)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 4-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™
CAS: 160388-54-5 Molecular Formula: C10H9N3O2 Molecular Weight (g/mol): 203.201 MDL Number: MFCD07187900 InChI Key: DJXUEFGFCMIONN-UHFFFAOYSA-N PubChem CID: 3159714 IUPAC Name: 4-(1,2,4-triazol-1-ylmethyl)benzoic acid SMILES: C1=CC(=CC=C1CN2C=NC=N2)C(=O)O