accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHO

CAS RN 160388-56-7

IUPAC Name: {4-[(1H-1,2,4-triazol-1-yl)methyl]phenyl}methanol
Synonyms: [4-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: XEOWXPICRQBFIT-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CN2C=NC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results