accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3PClONCH3H3C

CAS RN 1605-65-8

IUPAC Name: bis(dimethylamino)phosphinoyl chloride
Synonyms: didimethylaminophosphinoyl chloride
Molecular Weight (g/mol): 170.58
Molecular Formula: C4H12ClN2OP
InChi Key: WYLQARGYFXBZMD-UHFFFAOYSA-N
SMILES: CN(C)P(Cl)(=O)N(C)C
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)