CAS RN 16052-06-5
EPPS, 99%
CAS: 16052-06-5 Molecular Formula: C9H20N2O4S Molecular Weight (g/mol): 252.329 MDL Number: MFCD00006160 InChI Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N Synonym: hepps,epps,4-2-hydroxyethyl-1-piperazinepropanesulfonic acid,3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid,1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl,n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid,4-2-hydroxyethyl piperazine-1-propanesulfonic acid,4-2-hydroxyethyl-1-piperazinepropane sulfonic acid,4-2-hydroxyethyl-1-piperazinepropanesulphonic acid PubChem CID: 85255 ChEBI: CHEBI:42298 IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid SMILES: C1CN(CCN1CCCS(=O)(=O)O)CCO
Thermo Scientific Chemicals EPPS, 99%, for biochemistry
CAS: 16052-06-5 Molecular Formula: C9H20N2O4S Molecular Weight (g/mol): 252.329 MDL Number: MFCD00006160 InChI Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N Synonym: 3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid PubChem CID: 85255 ChEBI: CHEBI:42298 IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid SMILES: C1CN(CCN1CCCS(=O)(=O)O)CCO
Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.5
CAS: 16052-06-5 Molecular Formula: C9H20N2O4S Molecular Weight (g/mol): 252.329 MDL Number: MFCD00006160 InChI Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N Synonym: HEPPS PubChem CID: 85255 ChEBI: CHEBI:42298 IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid SMILES: C1CN(CCN1CCCS(=O)(=O)O)CCO
HEPPS, (Crystalline),MP Biomedicals
CAS: 16052-06-5 Molecular Formula: C9H20N2O4S Molecular Weight (g/mol): 252.329 InChI Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N Synonym: hepps,epps,4-2-hydroxyethyl-1-piperazinepropanesulfonic acid,3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid,1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl,n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid,4-2-hydroxyethyl piperazine-1-propanesulfonic acid,4-2-hydroxyethyl-1-piperazinepropane sulfonic acid,4-2-hydroxyethyl-1-piperazinepropanesulphonic acid PubChem CID: 85255 ChEBI: CHEBI:42298 IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid SMILES: C1CN(CCN1CCCS(=O)(=O)O)CCO
Thermo Scientific Chemicals EPPS, 0.2M buffer soln., pH 8.0
CAS: 16052-06-5 Molecular Formula: C9H20N2O4S Molecular Weight (g/mol): 252.329 MDL Number: MFCD00006160 InChI Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N Synonym: HEPPS PubChem CID: 85255 ChEBI: CHEBI:42298 IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid SMILES: C1CN(CCN1CCCS(=O)(=O)O)CCO
4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic Acid 98.0+%, TCI America™
CAS: 16052-06-5 Molecular Formula: C9H20N2O4S Molecular Weight (g/mol): 252.329 MDL Number: MFCD00006160 InChI Key: OWXMKDGYPWMGEB-UHFFFAOYSA-N Synonym: hepps,epps,4-2-hydroxyethyl-1-piperazinepropanesulfonic acid,3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid,1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl,n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid,4-2-hydroxyethyl piperazine-1-propanesulfonic acid,4-2-hydroxyethyl-1-piperazinepropane sulfonic acid,4-2-hydroxyethyl-1-piperazinepropanesulphonic acid PubChem CID: 85255 ChEBI: CHEBI:42298 IUPAC Name: 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid SMILES: C1CN(CCN1CCCS(=O)(=O)O)CCO