accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOSH2NHCl

CAS RN 160521-72-2

IUPAC Name: 1-{5-[(thiophen-2-yl)methoxy]-1H-indol-3-yl}propan-2-amine hydrochloride
Synonyms: 1-[5-(thiophen-2-ylmethoxy)-1H-indol-3-yl]propan-2-amine hydrochloride
Molecular Weight (g/mol): 322.85
Molecular Formula: C16H19ClN2OS
InChi Key: PYJBJMIBANAOFJ-UHFFFAOYNA-N
SMILES: Cl.CC(N)CC1=CNC2=CC=C(OCC3=CC=CS3)C=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results