accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHOFFF

CAS RN 160542-02-9

IUPAC Name: 1-ethynyl-4-(trifluoromethoxy)benzene
Synonyms: 1-ethynyl-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 186.13
Molecular Formula: C9H5F3O
InChi Key: RWWGGRCLMVYXPM-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)C#C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)