accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160591-91-3

IUPAC Name: (4-chloro-2-fluorophenyl)boronic acid
Synonyms: 4-chloro-2-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: YBNDRTRLXPEWKQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)C=C1F
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)