CAS RN 16064-08-7
IUPAC Name:
6-iodo-1,4-dihydroquinazolin-4-one
Synonyms:
6-iodo-1H-quinazolin-4-one
Molecular Weight (g/mol):
272.05
Molecular Formula:
C8H5IN2O
InChi Key:
PUGXMZKDRVGIHC-UHFFFAOYSA-N
SMILES:
IC1=CC=C2NC=NC(=O)C2=C1
1
–
1
of
1
results
1
6-Iodo-4-hydroxyquinazoline 97.0+%, TCI America™
CAS: 16064-08-7 Molecular Formula: C8H5IN2O Molecular Weight (g/mol): 272.045 MDL Number: MFCD00460007 InChI Key: PUGXMZKDRVGIHC-UHFFFAOYSA-N Synonym: 6-Iodo-4-quinazolinol, 6-Iodo-4-quinazolinone PubChem CID: 763741 IUPAC Name: 6-iodo-1H-quinazolin-4-one SMILES: C1=CC2=C(C=C1I)C(=O)N=CN2