accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16078-71-0

IUPAC Name: ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate
Synonyms: ethyl 5-amino-1-phenylpyrazole-4-carboxylate
Molecular Weight (g/mol): 231.26
Molecular Formula: C12H13N3O2
InChi Key: AYJIUOZKKTUKKD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)N(N=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results