CAS RN 160780-82-5
IUPAC Name:
9-{4-[4'-(9H-carbazol-9-yl)-5-[4-(9H-carbazol-9-yl)phenyl]-[1,1'-biphenyl]-3-yl]phenyl}-9H-carbazole
Synonyms:
9-{4-[4'-(carbazol-9-yl)-5-[4-(carbazol-9-yl)phenyl]-[1,1'-biphenyl]-3-yl]phenyl}carbazole
Molecular Weight (g/mol):
801.99
Molecular Formula:
C60H39N3
InChi Key:
XCICDYGIJBPNPC-UHFFFAOYSA-N
SMILES:
C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2