accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16092-38-9

IUPAC Name: bis(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) cobalt
Synonyms: bis(5,5,5-trifluoro-4-hydroxypent-3-en-2-one) cobalt
Molecular Weight (g/mol): 367.11
Molecular Formula: C10H10CoF6O4
InChi Key: GCHODJIKBYKCNY-UHFFFAOYSA-N
SMILES: [Co].CC(=O)C=C(O)C(F)(F)F.CC(=O)C=C(O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results