accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OOSCSSCSOOCH3CH3

CAS RN 1609536-17-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 753.15
Molecular Formula: C42H56O4S4
InChi Key: WZMUYZASTRRKIK-UHFFFAOYNA-N
SMILES: CCCCCCC(CCCC)COC(=O)C1=C(S[C]=C1)C1=CC=C(S1)C1=CC=C(S1)C1=C(C=[C]S1)C(=O)OCC(CCCC)CCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)