accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCOOOONHONOO

CAS RN 1609651-90-2

IUPAC Name: 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propanamide
Synonyms: 3-(2,5-dioxopyrrol-1-yl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propanamide
Molecular Weight (g/mol): 382.41
Molecular Formula: C18H26N2O7
InChi Key: GHEBECZRHJXPTL-UHFFFAOYSA-N
SMILES: O=C(CCN1C(=O)C=CC1=O)NCCOCCOCCOCCOCC#C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)