accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160982-10-5

IUPAC Name: 3-acetyl-5-chlorothiophene-2-sulfonamide
Synonyms: 3-acetyl-5-chlorothiophene-2-sulfonamide
Molecular Weight (g/mol): 239.69
Molecular Formula: C6H6ClNO3S2
InChi Key: ODLFFSHLXVZFPY-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(SC(Cl)=C1)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results