accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160982-16-1

IUPAC Name: 6-chloro-4-hydroxy-2H,3H,4H-1λ⁶-thieno[3,2-e][1,2]thiazine-1,1-dione
Synonyms: 6-chloro-4-hydroxy-2H,3H,4H-1λ⁶-thieno[3,2-e][1,2]thiazine-1,1-dione
Molecular Weight (g/mol): 239.69
Molecular Formula: C6H6ClNO3S2
InChi Key: OFJGKGNJDCLNPM-UHFFFAOYNA-N
SMILES: OC1CNS(=O)(=O)C2=C1C=C(Cl)S2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results