accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN

CAS RN 161082-75-3

IUPAC Name: 4-(dodecyloxy)benzene-1,2-dicarbonitrile
Synonyms: 4-(dodecyloxy)benzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 312.46
Molecular Formula: C20H28N2O
InChi Key: SWVGQFFVXRAJRQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC(C#N)=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results