accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1611-83-2

IUPAC Name: 2-methyl-N-phenylprop-2-enamide
Synonyms: 2-methyl-N-phenylprop-2-enamide
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: IJSVVICYGLOZHA-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results