CAS RN 161105-54-0
IUPAC Name:
(2S)-4-(tert-butoxy)-4-oxobutan-2-aminium
Synonyms:
(2S)-4-(tert-butoxy)-4-oxobutan-2-aminium
Molecular Weight (g/mol):
160.24
Molecular Formula:
C8H18NO2
InChi Key:
BFFNZGWJTHWUMY-LURJTMIESA-O
SMILES:
C[C@H]([NH3+])CC(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
tert-Butyl (3S)-3-aminobutanoate, 95%, Thermo Scientific Chemicals
tert-Butyl (3S)-3-aminobutanoate, 95%, Thermo Scientific Chemicals
CAS: 161105-54-0 MDL Number: MFCD00798294 InChI Key: BFFNZGWJTHWUMY-LURJTMIESA-N PubChem CID: 7021083 IUPAC Name: tert-butyl (3S)-3-aminobutanoate SMILES: CC(CC(=O)OC(C)(C)C)N