CAS RN 16111-27-6
IUPAC Name:
[2-(carbamimidoylsulfanyl)ethyl]dimethylamine hydrochloridyl
Synonyms:
[2-(carbamimidoylsulfanyl)ethyl]dimethylamine hydrochloridyl
Molecular Weight (g/mol):
182.69
Molecular Formula:
C5H13ClN3S
InChi Key:
NEQDKQTXUQVFFU-UHFFFAOYSA-N
SMILES:
[Cl].CN(C)CCSC(N)=N