accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16114-05-9

IUPAC Name: N'-{[(dimethylamino)methylidene]amino}-N,N-dimethylmethanimidamide
Synonyms: N'-{[(dimethylamino)methylidene]amino}-N,N-dimethylmethanimidamide
Molecular Weight (g/mol): 142.21
Molecular Formula: C6H14N4
InChi Key: AYTAYKXVFZPRAF-UHFFFAOYSA-N
SMILES: CN(C)C=NN=CN(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results