accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16114-47-9

IUPAC Name: (3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
Synonyms: 3,5-dimethyl-1,2-oxazol-4-ylboronic acid
Molecular Weight (g/mol): 140.93
Molecular Formula: C5H8BNO3
InChi Key: DIIFZCPZIRQDIJ-UHFFFAOYSA-N
SMILES: CC1=C(B(O)O)C(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results