accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3Br

CAS RN 16116-78-2

IUPAC Name: [2-(4-bromophenyl)ethynyl]trimethylsilane
Synonyms: [2-(4-bromophenyl)ethynyl]trimethylsilane
Molecular Weight (g/mol): 253.21
Molecular Formula: C11H13BrSi
InChi Key: RNMSGCJGNJYDNS-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results