accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS(S)ONH2NH2HCl

CAS RN 16120-92-6

IUPAC Name: 2-amino-4-(methylsulfanyl)butanamide hydrochloride
Synonyms: 2-amino-4-(methylsulfanyl)butanamide hydrochloride
Molecular Weight (g/mol): 184.68
Molecular Formula: C5H13ClN2OS
InChi Key: PWCBMJHINDTXGV-UHFFFAOYNA-N
SMILES: Cl.CSCCC(N)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results