accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 161233-97-2

NOHON

CAS RN 161233-97-2

IUPAC Name: 2-cyanopyridine-4-carboxylic acid
Synonyms: 2-cyanopyridine-4-carboxylic acid
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: MPSVJNPESHZCIB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results