accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HSHS

CAS RN 16128-68-0

IUPAC Name: butane-1,2-dithiol
Synonyms: butane-1,2-dithiol
Molecular Weight (g/mol): 122.24
Molecular Formula: C4H10S2
InChi Key: LFTMJBWNOFFSRW-UHFFFAOYNA-N
SMILES: CCC(S)CS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results