accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOOCl

CAS RN 16133-25-8

IUPAC Name: pyridine-3-sulfonyl chloride
Synonyms: pyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 177.6
Molecular Formula: C5H4ClNO2S
InChi Key: CDRNYKLYADJTMN-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results