accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16133-49-6

IUPAC Name: 5-methoxy-2-nitroaniline
Synonyms: 3-amino-4-nitro-anisol
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: QEHVRGACCVLLNN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(N)=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)