accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BOH3CH3CH3CCH3O(E)

CAS RN 161395-96-6

IUPAC Name: 4,4,5,5-tetramethyl-2-[(1E)-pent-1-en-1-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[(1E)-pent-1-en-1-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 196.1
Molecular Formula: C11H21BO2
InChi Key: IMIHEZMTQOARIK-CMDGGOBGSA-N
SMILES: CCC\C=C\B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)