accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNNN

CAS RN 1614-12-6

IUPAC Name: 1H-1,2,3-benzotriazol-1-amine
Synonyms: 1,2,3-benzotriazol-1-amine
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: JCXKHYLLVKZPKE-UHFFFAOYSA-N
SMILES: NN1N=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)