CAS RN 161468-32-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 4-Cyano-3-methylisoquinoline
A potent, cell-permeable, reversible, and specific inhibitor of protein kinase A (IC50 = 30 nM).