accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONHO

CAS RN 161511-90-6

IUPAC Name: tert-butyl 2-(hydroxymethyl)azetidine-1-carboxylate
Synonyms: tert-butyl 2-(hydroxymethyl)azetidine-1-carboxylate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: XIRUXUKRGUFEKC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC1CO
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)