accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFFF

CAS RN 161622-05-5

IUPAC Name: 3-fluoro-5-(trifluoromethyl)benzoate
Synonyms: 3-fluoro-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 207.1
Molecular Formula: C8H3F4O2
InChi Key: NSGKIIGVPBTOBF-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results