accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 161712-75-0

OHOFF

CAS RN 161712-75-0

IUPAC Name: 3-(3,4-difluorophenyl)propanoic acid
Synonyms: 3-(3,4-difluorophenyl)propanoic acid
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: UOZIYCHJMUNLIG-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results