accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16184-89-7

IUPAC Name: 1-(4-bromophenyl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(4-bromophenyl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 253.02
Molecular Formula: C8H4BrF3O
InChi Key: IHGSAQHSAGRWNI-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=CC=C(Br)C=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results