accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOHO

CAS RN 16188-55-9

IUPAC Name: 2-[4-(methylsulfanyl)phenyl]acetic acid
Synonyms: [4-(methylsulfanyl)phenyl]acetic acid
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: AHMLFHMRRBJCRM-UHFFFAOYSA-N
SMILES: CSC1=CC=C(CC(O)=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)