accessibility menu, dialog, popup

UserName

Viewing profile as user:
HON

CAS RN 1619-34-7

IUPAC Name: 1-azabicyclo[2.2.2]octan-3-ol
Synonyms: 1-azabicyclo 2.2.2 octan-3-ol
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: IVLICPVPXWEGCA-UHFFFAOYNA-N
SMILES: OC1CN2CCC1CC2
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)