accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1619-58-5

IUPAC Name: ethyl 2-cyanobutanoate
Synonyms: ethyl 2-cyanobutanoate
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: FCISHUHNFYJJDU-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CC)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results