accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 161957-56-8

IUPAC Name: 3-bromo-2-fluorobenzoic acid
Synonyms: 3-bromo-2-fluorobenzoic acid
Molecular Weight (g/mol): 219.01
Molecular Formula: C7H4BrFO2
InChi Key: UVKURTLVTLRSSM-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(Br)=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)