accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1620-75-3

IUPAC Name: 6-methylpyridine-2-carbonitrile
Synonyms: 2-cyano-6-methylpyridine
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: CMADFEQMYFNYCF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=N1)C#N
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)