accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOOHO

CAS RN 162046-56-2

IUPAC Name: 4-({1-[(tert-butoxy)carbonyl]piperidin-4-yl}oxy)benzoic acid
Synonyms: 4-{[1-(tert-butoxycarbonyl)piperidin-4-yl]oxy}benzoic acid
Molecular Weight (g/mol): 321.37
Molecular Formula: C17H23NO5
InChi Key: LUVXDMFUUZGFST-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results