accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16205-84-8

IUPAC Name: ethyl 3-(trimethylsilyl)prop-2-ynoate
Synonyms: ethyl 3-(trimethylsilyl)prop-2-ynoate
Molecular Weight (g/mol): 170.28
Molecular Formula: C8H14O2Si
InChi Key: QRBKPHYAIRLCLK-UHFFFAOYSA-N
SMILES: CCOC(=O)C#C[Si](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)