accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1621-91-6

IUPAC Name: 1H-pyrazole-5-carboxylic acid
Synonyms: 2H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: KOPFEFZSAMLEHK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NN1
Quantity 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results