accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NNCH3H3CH3CBrBr(E/Z)(E/Z)(E/Z)

CAS RN 162112-35-8

IUPAC Name: 4-{6-[4-(diethylamino)phenyl]hexa-1,3,5-trien-1-yl}-1-[3-(triethylazaniumyl)propyl]pyridin-1-ium dibromide
Synonyms: 4-{6-[4-(diethylamino)phenyl]hexa-1,3,5-trien-1-yl}-1-[3-(triethylammonio)propyl]pyridin-1-ium dibromide
Molecular Weight (g/mol): 607.52
Molecular Formula: C30H45Br2N3
InChi Key: AFVSZGYRRUMOFH-UHFFFAOYSA-L
SMILES: [Br-].[Br-].CCN(CC)C1=CC=C(C=CC=CC=CC2=CC=[N+](CCC[N+](CC)(CC)CC)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results