accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3N

CAS RN 16213-85-7

IUPAC Name: 2-(2,5-dimethylphenyl)acetonitrile
Synonyms: acetonitrile, (2,5-xylyl)-
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: HCELORQTHSKCQL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(CC#N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results