accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 162148-48-3

H3CH3CCH3OONHNNNHOOCH3H3CCH3

CAS RN 162148-48-3

IUPAC Name: tert-butyl 2-{7-[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Synonyms: tert-butyl 2-{7-[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Molecular Weight (g/mol): 400.56
Molecular Formula: C20H40N4O4
InChi Key: OERWIIJOLSXGAQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results